2-Fluoro-6-iodoaniline 97%

Supplier: Ambeed
Warning

A110636-1G A110636-250MG A110636-5G
77257-640EA 84.46 USD
77257-640 77257-642 77257-644
2-Fluoro-6-iodoaniline 97%
2-Fluoro-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.013 g/mol
MDL Number: MFCD07368681
CAS Number: 886762-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR