Methyl-3-bromo-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A110620-100G A110620-10G A110620-1G A110620-25G A110620-500G A110620-5G
77270-880EA 75 USD
77270-880 77270-882 77270-884 77270-886 77270-888 77270-890
Methyl-3-bromo-2-methylbenzoate 98%
Methyl 3-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD09031772
CAS Number: 99548-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR