4'-Amino-3',5'-dichloroacetophenone 98%

Supplier: Ambeed
Warning

A110431-10G A110431-1G A110431-5G A110431-100G A110431-25G A110431-500G
77233-518EA 11 USD
77233-518 77233-520 77233-522 77614-850 77745-820 77745-821
4'-Amino-3',5'-dichloroacetophenone 98%
4'-Amino-3',5'-dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
MDL Number: MFCD00238535
CAS Number: 37148-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR