4,4'-(Hexafluoroisopropylidene)diphthalic anhydride 98%

Supplier: Ambeed
Danger

Synonyms: 5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane

A110372-100G A110372-10G A110372-1G A110372-25G A110372-500G A110372-5G
77283-988EA 63 USD
77283-988 77283-990 77283-992 77283-994 77283-996 77283-998
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride 98%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₄O₃
MDL Number: MFCD00039143
CAS Number: 1107-00-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR