4-Methyl-3-nitrophenylacetic acid 98%

Supplier: Ambeed
Warning

A110350-1G A110350-250MG A110350-5G
77283-982EA 7 USD
77283-982 77283-984 77283-986
4-Methyl-3-nitrophenylacetic acid 98%
4-Methyl-3-nitrophenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD09832994
CAS Number: 54941-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR