Trimethyl pyridine-2,4,6-tricarboxylate 95%

Supplier: Ambeed
Warning

A110347-100MG A110347-1G A110347-250MG
77233-506EA 25 USD
77233-506 77233-508 77233-510
Trimethyl pyridine-2,4,6-tricarboxylate 95%
Trimethyl pyridine-2,4,6-tricarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₆
MDL Number: MFCD26792768
CAS Number: 25309-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR