4-Oxo-4-((5-sulfamoyl-1,3,4-thiadiazol-2-yl)amino)butanoic acid 97%

Supplier: Ambeed
Warning

A110298-100MG A110298-1G A110298-250MG
77283-966EA 158.62 USD
77283-966 77283-968 77283-970
4-Oxo-4-((5-sulfamoyl-1,3,4-thiadiazol-2-yl)amino)butanoic acid 97%
4-Oxo-4-((5-sulfamoyl-1,3,4-thiadiazol-2-yl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₆H₈N₄O₅S₂
MW: 280.28 g/mol
MDL Number: MFCD01104616
CAS Number: 78851-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR