1-(2-Cyanophenyl)piperidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile

A110212-1G A110212-25G
77270-830EA 11.33 USD
77270-830 77270-832
1-(2-Cyanophenyl)piperidine 98%
1-(2-Cyanophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MW: 186.26 g/mol
MDL Number: MFCD00049221
CAS Number: 72752-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR