tert-Butylsulfonamide 98%

Supplier: Ambeed
Warning

A110151-100G A110151-1G A110151-25G A110151-5G
77283-944EA 249 USD
77283-944 77283-946 77283-948 77283-950
tert-Butylsulfonamide 98%
tert-Butylsulfonamide

Precaución: for Research Use Only

Formula: C₄H₁₁NO₂S
MDL Number: MFCD02179404
CAS Number: 34813-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR