3-Bromo-2,6-dimethylbenzoic acid 95%

Supplier: Ambeed
Warning

A110122-1G A110122-250MG A110122-5G
77245-654EA 66.95 USD
77245-654 77245-656 77592-178
3-Bromo-2,6-dimethylbenzoic acid 95%
3-Bromo-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD09260824
CAS Number: 123278-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR