5-tert-Butyl-L-glutamate hydrate 98%

Supplier: Ambeed
Warning

Synonyms: H-Glu(OtBu)-OH, L-Glutamic Acid 5-tert-Butyl Ester

A110082-1G A110082-25G A110082-5G A110082-100G A110082-10G A110082-500G
77257-618EA 6.18 USD
77257-618 77257-620 77257-622 77570-564 77570-566 77614-840
5-tert-Butyl-L-glutamate hydrate 98%
5-tert-Butyl-L-glutamate hydrate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD00038580
CAS Number: 2419-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR