KN-93 phosphate 97%

Supplier: Ambeed
Warning

A109921-10MG A109921-1MG A109921-25MG A109921-5MG A109921-50MG
77257-610EA 32 USD
77257-610 77257-612 77257-614 77257-616 77747-810
KN-93 phosphate 97%
KN-93 phosphate

Precaución: for Research Use Only

Formula: C₂₆H₂₉ClN₂O₄S·H₃PO₄
MDL Number: MFCD03788201
CAS Number: 1188890-41-6 | 139298-40-1 (free base)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR