5-Amino-2-chloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine

A109918-100G A109918-10G A109918-25G A109918-500G A109918-5G
77270-810EA 41.2 USD
77270-810 77270-812 77270-814 77270-816 77270-818
5-Amino-2-chloropyridine 98%
5-Amino-2-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD00006243
CAS Number: 5350-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR