2-Acetylcyclohexanone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetyl-1-cyclohexanone

A109905-100G A109905-10G A109905-1G A109905-25G A109905-5G A109905-500G
77245-638EA 86 USD
77245-638 77245-640 77245-642 77245-644 77245-646 77745-818
2-Acetylcyclohexanone 98%
2-Acetylcyclohexanone

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00001633
CAS Number: 874-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR