3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

A109836-10G A109836-1G A109836-5G
77283-894EA 80 USD
77283-894 77283-896 77283-898
3-(Morpholinomethyl)phenylboronic acid pinacol ester 97%
3-(Morpholinomethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₃
MW: 303.20 g/mol
MDL Number: MFCD06797489
CAS Number: 364794-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR