3-Methoxypropiononitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methoxypropanenitrile, 3-Methoxypropionitrile
A109791-100G
A109791-25G
A109791-500G
77270-796EA
8.24
USD
77270-796
77270-798
77270-800
3-Methoxypropiononitrile 98%
3-Methoxypropiononitrile
Precaución: for Research Use Only
Formula:
C₄H₇NO MW: 85.11 g/mol |
MDL Number:
MFCD00001958 CAS Number: 110-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...