1-Hydroxycyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A109621-10G A109621-1G A109621-250MG A109621-25G A109621-5G
77233-486EA 160.68 USD
77233-486 77233-488 77233-490 77233-492 77233-494
1-Hydroxycyclohexanecarboxylic acid 98%
1-Hydroxycyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00045477
CAS Number: 1123-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR