3-Methylpiperidin-2-one 95%

Supplier: Ambeed
Warning

A109558-100MG A109558-1G A109558-250MG A109558-5G
77270-772EA 12 USD
77270-772 77270-774 77270-776 77270-778
3-Methylpiperidin-2-one 95%
3-Methylpiperidin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MDL Number: MFCD00177941
CAS Number: 3768-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR