Ir[dF(CF3)ppy]2(dtbbpy)PF6 98%

Supplier: Ambeed
Warning

A109526-100MG A109526-1G A109526-250MG A109526-50MG A109526-5G
77270-752EA 21 USD
77270-752 77270-754 77270-756 77270-758 77270-760
Ir[dF(CF3)ppy]2(dtbbpy)PF6 98%
Ir[dF(CF3)ppy]2(dtbbpy)PF6

Precaución: for Research Use Only

Formula: C₄₂H₃₄F₁₆IrN₄P
MDL Number: MFCD27952556
CAS Number: 870987-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR