Indole-5-carbaldehyd 98%

Supplier: Ambeed
Warning

A109501-100G A109501-10G A109501-1G A109501-25G A109501-5G
77257-596EA 287 USD
77257-596 77257-598 77257-600 77257-602 77257-604
Indole-5-carbaldehyd 98%
Indole-5-carbaldehyd

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD02093664
CAS Number: 1196-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR