(4-Hydroxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenylacetic Acid

A109495-1000G A109495-100G A109495-100MG A109495-25G A109495-25MG A109495-500G
77245-622EA 63.86 USD
77245-622 77245-624 77245-626 77245-628 77245-630 77245-632
(4-Hydroxyphenyl)acetic acid 98%
(4-Hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
MDL Number: MFCD00004347
CAS Number: 156-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR