(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

A109453-100MG A109453-1G A109453-250MG A109453-5G
77245-614EA 35.02 USD
77245-614 77245-616 77245-618 77245-620
(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)ethanol 97%
(S)-2-Amino-2-(3-ethoxy-4-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.25 g/mol
MDL Number: MFCD09253802
CAS Number: 1213876-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR