3-Fluorobenzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A109337-100G A109337-1G A109337-250MG A109337-25G A109337-5G
77245-604EA 754.99 USD
77245-604 77245-606 77245-608 77245-610 77245-612
3-Fluorobenzene-1,2-diamine 98%
3-Fluorobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C6H7FN2
MDL Number: MFCD00061130
CAS Number: 18645-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR