2-Chloro-5-fluoroaniline 98%

Supplier: Ambeed
Warning

A109284-10G A109284-5G A109284-100G A109284-25G
77283-852EA 19.57 USD
77283-852 77283-854 77549-104 77614-834
2-Chloro-5-fluoroaniline 98%
2-Chloro-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD00084852
CAS Number: 452-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR