5-Chloro-2-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A109198-100G A109198-10G A109198-1G A109198-25G
77233-424EA 49 USD
77233-424 77233-426 77233-428 77233-430 77233-432
5-Chloro-2-methoxybenzoic acid 95%

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD00017546
CAS Number: 3438-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR