4-Bromo-3,5-dimethylanisole 98%

Supplier: Ambeed
Warning

A109175-100G A109175-10G A109175-25G A109175-500G A109175-5G
77283-834EA 161 USD
77283-834 77283-836 77283-838 77283-840 77283-842 77283-844
4-Bromo-3,5-dimethylanisole 98%
4-Bromo-3,5-dimethylanisole

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD00461692
CAS Number: 6267-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR