2-Chloro-4-iodo-6-nitroaniline 97%
Supplier: Ambeed
|
Warning
|
A109106-100MG
A109106-10G
A109106-1G
A109106-250MG
A109106-5G
77233-414EA
5
USD
77233-414
77233-416
77233-418
77233-420
77233-422
2-Chloro-4-iodo-6-nitroaniline 97%
2-Chloro-4-iodo-6-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄ClIN₂O₂ |
MDL Number:
MFCD26936371 CAS Number: 873386-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...