2-Chloro-6-methylquinoline 98%

Supplier: Ambeed
Warning

A109105-1G A109105-25G A109105-5G
77257-578EA 246.17 USD
77257-578 77257-580 77257-582 77257-584
2-Chloro-6-methylquinoline 98%
2-Chloro-6-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MDL Number: MFCD00462389
CAS Number: 4295-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR