1,2,3-Benzenetricarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-Benzenetricarboxylic acid, Benzene-1,2,3-tricarboxylic acid, Hemimellitic acid

A109046-1G A109046-25G A109046-5G
77257-574EA 9.27 USD
77257-574 77257-576 77570-522
1,2,3-Benzenetricarboxylic acid 95%
1,2,3-Benzenetricarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₆
MW: 210.14 g/mol
MDL Number: MFCD00002468
CAS Number: 569-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR