4-Cyclopropyl-benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyclopropylbenzoic acid

A108884-250MG A108884-1G A108884-100MG
77283-788EA 30 USD
77283-788 77570-510 77747-805
4-Cyclopropyl-benzoic acid 97%
4-Cyclopropyl-benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD01069076
CAS Number: 1798-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR