2-(5-Methylfuran-2-yl)ethanol 97%

Supplier: Ambeed
Warning

A108861-1G A108861-250MG
77257-570EA 121 USD
77257-570 77257-572
2-(5-Methylfuran-2-yl)ethanol 97%
2-(5-Methylfuran-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MDL Number: MFCD15145904
CAS Number: 35942-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR