Dibenzo[b,f][1,5]diazocine-6,12(5H,11H)-dione 97%

Supplier: Ambeed
Warning

A108854-1G A108854-250MG A108854-5G
77257-564EA 71.07 USD
77257-564 77257-566 77257-568
Dibenzo[b,f][1,5]diazocine-6,12(5H,11H)-dione 97%
Dibenzo[b,f][1,5]diazocine-6,12(5H,11H)-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₂
MW: 238.24 g/mol
MDL Number: MFCD00655993
CAS Number: 15351-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR