2-(Methylthio)-2-thiazoline 98%

Supplier: Ambeed
Warning

A108788-1G A108788-25G A108788-5G
77245-594EA 9 USD
77245-594 77245-596 77245-598
2-(Methylthio)-2-thiazoline 98%
2-(Methylthio)-2-thiazoline

Precaución: for Research Use Only

Formula: C₄H₇NS₂
MDL Number: MFCD00005312
CAS Number: 19975-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR