6-Amino-2-methoxynicotinic acid 97%

Supplier: Ambeed
Warning

A108783-1G A108783-250MG
77245-590EA 276 USD
77245-590 77245-592
6-Amino-2-methoxynicotinic acid 97%
6-Amino-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD08062960
CAS Number: 1060806-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR