Piperonyl butoxide 95+%

Supplier: Ambeed
Warning

Synonyms: 5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401

A108763-1000G A108763-100G A108763-25G A108763-500G
77233-380EA 127 USD
77233-380 77233-382 77233-384 77233-386
Piperonyl butoxide 95+%
Piperonyl butoxide

Precaución: for Research Use Only

Formula: C₁₉H₃₀O₅
MDL Number: MFCD00005842
CAS Number: 51-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR