(6-Bromoimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone 97%

Supplier: Ambeed
Warning

A108714-1G A108714-250MG
77257-558EA 13.39 USD
77257-558 77257-560
(6-Bromoimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone 97%
(6-Bromoimidazo[1,2-a]pyridin-3-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₉BrN₂O
MW: 301.13 g/mol
MDL Number: MFCD29904976
CAS Number: 1284293-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR