Dichloridebis(1,10-phenanthroline)(2,2'-bipyridine)ruthenium(II) 98%

Supplier: Ambeed
Warning

A108702-100MG A108702-250MG
77257-554EA 63 USD
77257-554 77257-556
Dichloridebis(1,10-phenanthroline)(2,2'-bipyridine)ruthenium(II) 98%
Dichloridebis(1,10-phenanthroline)(2,2'-bipyridine)ruthenium(II)

Precaución: for Research Use Only

Formula: C₃₄H₂₄ClN₆Ru
MW: 653.11 g/mol
CAS Number: 60828-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR