8-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one 97%

Supplier: Ambeed
Warning

A108675-10MG A108675-1MG A108675-25MG A108675-5MG
77245-580EA 50.47 USD
77245-580 77245-582 77245-584 77245-586
8-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one 97%
8-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₆
MW: 610.52 g/mol
CAS Number: 76135-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR