β-D-Ribofuranose-1,2,3,5-tetraacetate 97%

Supplier: Ambeed
Warning

Synonyms: Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

A108665-1000G A108665-100G A108665-25G A108665-500G A108665-5G
77270-714EA 29 USD
77270-714 77270-716 77270-718 77270-720 77270-722
β-D-Ribofuranose-1,2,3,5-tetraacetate 97%
beta-D-Ribofuranose-1,2,3,5-tetraacetate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₉
MDL Number: MFCD00005358
CAS Number: 13035-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR