(3,4-Epoxycyclohexyl)methyl acrylate 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with hydroquinone (HQ)

A108658-100G A108658-10G A108658-1G A108658-25G A108658-5G
77233-364EA 55 USD
77233-364 77233-366 77233-368 77233-370 77233-372
(3,4-Epoxycyclohexyl)methyl acrylate 95% stabilized
(3,4-Epoxycyclohexyl)methyl acrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD28364662
CAS Number: 64630-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR