Benzyl morpholine-4-carboxylate 95%

Supplier: Ambeed
Warning

A108639-250MG A108639-1G A108639-5G
77245-578EA 17.51 USD
77245-578 77614-828 77745-810
Benzyl morpholine-4-carboxylate 95%
Benzyl morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.25 g/mol
MDL Number: MFCD11111783
CAS Number: 25070-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR