CYM 5541 98%

Supplier: Ambeed
Warning

Synonyms: CYM-5541, N,N-Dicyclohexyl-5-cyclopropylisoxazole-3-carboxamide

A108628-10MG A108628-1MG A108628-25MG A108628-50MG A108628-5MG
77270-704EA 66 USD
77270-704 77270-706 77270-708 77270-710 77270-712
CYM 5541 98%
CYM 5541

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₂O₂
MW: 316.43 g/mol
MDL Number: MFCD09782785
CAS Number: 945128-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR