(3-Iodophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Iodophenyl)acetic acid

A108605-100G A108605-10G A108605-1G A108605-250MG A108605-25G A108605-5G A108605-100MG
77233-352EA 432 USD
77233-352 77233-354 77233-356 77233-358 77233-360 77233-362 77570-498
(3-Iodophenyl)acetic acid 98%
(3-Iodophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄I)CH₂COOH
MDL Number: MFCD00046548
CAS Number: 1878-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR