2-Fluororesorcinol 97%

Supplier: Ambeed
Warning

A108591-100MG A108591-1G A108591-250MG
77257-548EA 39 USD
77257-548 77257-550 77257-552
2-Fluororesorcinol 97%
2-Fluororesorcinol

Precaución: for Research Use Only

Formula: C₆H₅FO₂
MDL Number: MFCD09835187
CAS Number: 103068-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR