RSL3 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3R)-2-(2-Chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1H-pyrido[3,4-b]indole-3-carboxylic acid methyl ester, (1S,3R)-Methyl 2-(2-chloroacetyl)-1-(4-(methoxycarbonyl)phenyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate

A108537-100MG A108537-250MG A108537-50MG A108537-1G A108537-1MG A108537-2MG
77233-344EA 172 USD
77233-344 77233-346 77233-348 77745-807 77745-808 77745-809
RSL3 98%
RSL3

Precaución: for Research Use Only

Formula: C₂₃H₂₁ClN₂O₅
MDL Number: MFCD30187526
CAS Number: 1219810-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR