4,6-Dichloro-5-methylpyrimidine 98%

Supplier: Ambeed
Danger

A108407-1G A108407-25G A108407-5G
77283-728EA 6.18 USD
77283-728 77283-730 77283-732
4,6-Dichloro-5-methylpyrimidine 98%
4,6-Dichloro-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD01646112
CAS Number: 4316-97-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR