2-Chloro-6-nitrophenol 97%

Supplier: Ambeed
Danger

A108391-10G A108391-1G A108391-250MG A108391-5G
77283-720EA 27 USD
77283-720 77283-722 77283-724 77283-726
2-Chloro-6-nitrophenol 97%
2-Chloro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD00024233
CAS Number: 603-86-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR