cis-3,6-Bis(2-hydroxyethyl)piperazine-2,5-dione 97%

Supplier: Ambeed
Warning

A108363-10G A108363-1G A108363-250MG
77233-342EA 219.39 USD
77233-342 77614-824 77745-806
cis-3,6-Bis(2-hydroxyethyl)piperazine-2,5-dione 97%
cis-3,6-Bis(2-hydroxyethyl)piperazine-2,5-dione

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
MDL Number: MFCD03788820
CAS Number: 28814-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR