MK-5108 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(3-chloro-2-fluorophenoxy)-1-[[6-(1,3-thiazol-2-ylamino)pyridin-2-yl]methyl]cyclohexane-1-carboxylic acid, trans-4-(3-Chloro-2-fluorophenoxy)-1-((6-(thiazol-2-ylamino)pyridin-2-yl)methyl)cyclohexane-1-carboxylic acid
A108356-100MG
A108356-10MG
A108356-1MG
A108356-5MG
77233-334EA
536
USD
77233-334
77233-336
77233-338
77233-340
MK-5108 98%
MK-5108
Precaución: for Research Use Only
Formula:
C₂₂H₂₁ClFN₃O₃S |
MDL Number:
MFCD22124479 CAS Number: 1010085-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...