3-Fluoro-5-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A108353-1G A108353-5G A108353-10G
77270-694EA 11 USD
77270-694 77270-696 77747-799
3-Fluoro-5-nitrobenzoic acid 98%
3-Fluoro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MDL Number: MFCD05864360
CAS Number: 14027-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR